accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13709-31-4

FHVFHFHO

CAS RN 13709-31-4

IUPAC Name: vanadium(V) oxytrifluoride
Synonyms: vanadium(V) oxytrifluoride
Molecular Weight (g/mol): 126.96
Molecular Formula: F3H3OV
InChi Key: HHQFLEDKAVLHOM-UHFFFAOYSA-N
SMILES: F.F.F.O=[V]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results