accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NaOVOO

CAS RN 13718-26-8

IUPAC Name: sodium vanadiumoylolate
Synonyms: sodium metavanadate
Molecular Weight (g/mol): 121.93
Molecular Formula: NaO3V
InChi Key: CMZUMMUJMWNLFH-UHFFFAOYSA-N
SMILES: [Na+].[O-][V](=O)=O
Grade 
  • (5)
Physical Form 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)