accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13734-28-6

CH3H3CCH3OONH(S)H2NHOO

CAS RN 13734-28-6

IUPAC Name: 6-amino-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms: 6-amino-2-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol): 246.31
Molecular Formula: C11H22N2O4
InChi Key: DQUHYEDEGRNAFO-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCCCN)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)