accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SiH3CCH3OCH2

CAS RN 13735-81-4

IUPAC Name: trimethyl[(1-phenylethenyl)oxy]silane
Synonyms: trimethyl[(1-phenylethenyl)oxy]silane
Molecular Weight (g/mol): 192.33
Molecular Formula: C11H16OSi
InChi Key: AFFPCIMDERUIST-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC(=C)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results