accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOOCH3OOOHO

CAS RN 13739-02-1

IUPAC Name: 4,5-bis(acetyloxy)-9,10-dioxo-9,10-dihydroanthracene-2-carboxylic acid
Synonyms: diacerein artrodar diacereine diacereina rhein, diacetate (6CI) diacetyl-rhein niverpharm brand of diacetylrhein diacerhein art brand of diacetylrhein diacetylrhein
Molecular Weight (g/mol): 368.3
Molecular Formula: C19H12O8
InChi Key: TYNLGDBUJLVSMA-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC2=C1C(=O)C1=C(OC(C)=O)C=C(C=C1C2=O)C(O)=O
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results