accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OFOClF

CAS RN 1373920-79-6

IUPAC Name: 4-ethoxy-2,6-difluorobenzoyl chloride
Synonyms: 4-ethoxy-2,6-difluorobenzoyl chloride
Molecular Weight (g/mol): 220.6
Molecular Formula: C9H7ClF2O2
InChi Key: DFCDPQTYYGJYOD-UHFFFAOYSA-N
SMILES: CCOC1=CC(F)=C(C(Cl)=O)C(F)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results