accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COFFFFO

CAS RN 1373921-01-7

IUPAC Name: 3-fluoro-6-methoxy-2-(trifluoromethyl)benzaldehyde
Synonyms: 3-fluoro-6-methoxy-2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 222.14
Molecular Formula: C9H6F4O2
InChi Key: AYOBDUXUFBKTIY-UHFFFAOYSA-N
SMILES: COC1=C(C=O)C(=C(F)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results