accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13746-96-8

NOOONOOONOOONd

CAS RN 13746-96-8

IUPAC Name: neodymium(3+) hydrate trinitrate
Synonyms: neodymium(3+) hydrate trinitrate
Molecular Weight (g/mol): 348.27
Molecular Formula: H2N3NdO10
InChi Key: GZIAUVGSMHERLN-UHFFFAOYSA-N
SMILES: O.[Nd+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)