CAS RN 13750-68-0
IUPAC Name:
4-methyl-1,3-thiazole-2-carbaldehyde
Synonyms:
4-methyl-1,3-thiazole-2-carbaldehyde
Molecular Weight (g/mol):
127.16
Molecular Formula:
C5H5NOS
InChi Key:
NYMCQLLAIMUVSY-UHFFFAOYSA-N
SMILES:
CC1=CSC(C=O)=N1
1
–
1
of
1
results
1
4-Methyl-2-thiazolecarboxaldehyde, 97%
CAS: 13750-68-0 Molecular Formula: C5H5NOS Molecular Weight (g/mol): 127.17 MDL Number: MFCD00022450 InChI Key: NYMCQLLAIMUVSY-UHFFFAOYSA-N Synonym: 4-methylthiazole-2-carbaldehyde,4-methyl-2-thiazolecarboxaldehyde,4-methyl-thiazole-2-carbaldehyde,4-methyl-1,3-thiazole-2-carboxaldehyde,2-thiazolecarboxaldehyde, 4-methyl,4-methylthiazole-2-carboxaldehyde,4-methyl-2-formylthiazole,2-formyl-4-methyl-1,3-thiazole,4-methyl-2-thiazole carboxaldehyde,pubchem15253 PubChem CID: 4263432 IUPAC Name: 4-methyl-1,3-thiazole-2-carbaldehyde SMILES: CC1=CSC(=N1)C=O