CAS RN 13754-38-6
IUPAC Name:
4-benzoylpiperazin-1-ium
Synonyms:
4-benzoylpiperazin-1-ium
Molecular Weight (g/mol):
191.25
Molecular Formula:
C11H15N2O
InChi Key:
VUNXBQRNMNVUMV-UHFFFAOYSA-O
SMILES:
O=C(N1CC[NH2+]CC1)C1=CC=CC=C1
1
–
1
of
1
results
1
1-Benzoylpiperazine, 97%
CAS: 13754-38-6 Molecular Formula: C11H14N2O Molecular Weight (g/mol): 190.246 MDL Number: MFCD00810192 InChI Key: VUNXBQRNMNVUMV-UHFFFAOYSA-N Synonym: 1-benzoylpiperazine,n-benzoylpiperazine,phenyl-piperazin-1-yl-methanone,1-benzoyl piperazine,1-benzoyl-piperazine,phenyl piperazin-1-yl methanone,piperazine, 1-benzoyl,phenyl piperazinyl ketone,phenyl piperazin-4-ium-1-yl methanone,1-phenylcarbonyl piperazine PubChem CID: 762654 IUPAC Name: phenyl(piperazin-1-yl)methanone SMILES: C1CN(CCN1)C(=O)C2=CC=CC=C2