accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13760-80-0

IUPAC Name: ytterbium(3+) trifluoride
Synonyms: ytterbium(III) fluoride
Molecular Weight (g/mol): 230.05
Molecular Formula: F3Yb
InChi Key: XASAPYQVQBKMIN-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Yb+3]
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (5)
  • (2)
Quantity 
  • (1)
  • (3)
  • (3)
Grade 
  • (5)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results