accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13763-62-7

IUPAC Name: borane
Synonyms: borane boranediyl (singlet) boranediyl (triplet) borane(3) boron trihydride
Molecular Weight (g/mol): 13.83
Molecular Formula: BH3
InChi Key: UORVGPXVDQYIDP-UHFFFAOYSA-N
SMILES: B
Quantity 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results