accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3CH3CH3

CAS RN 13764-18-6

IUPAC Name: 1,4,6,7-tetramethylnaphthalene
Synonyms: 1,4,6,7-tetramethylnaphthalene
Molecular Weight (g/mol): 184.28
Molecular Formula: C14H16
InChi Key: VPSPONOBLZCLIU-UHFFFAOYSA-N
SMILES: CC1=CC2=C(C)C=CC(C)=C2C=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results