accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NBrBrF

CAS RN 137718-84-4

IUPAC Name: 3,4-dibromo-2-fluoropyridine
Synonyms: 3,4-dibromo-2-fluoropyridine
Molecular Weight (g/mol): 254.88
Molecular Formula: C5H2Br2FN
InChi Key: RALUGCFMNLTFAK-UHFFFAOYSA-N
SMILES: FC1=NC=CC(Br)=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results