accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13778-31-9

NOOONOOOCu

CAS RN 13778-31-9

IUPAC Name: copper(2+) hydrate dinitrate
Synonyms: copper(2+) hydrate dinitrate
Molecular Weight (g/mol): 205.57
Molecular Formula: CuH2N2O7
InChi Key: VMKYLARTXWTBPI-UHFFFAOYSA-N
SMILES: O.[Cu++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)