accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ONOSOOCl

CAS RN 1378261-28-9

IUPAC Name: 4-{1,3-dioxo-2-azaspiro[4.4]nonan-2-yl}benzene-1-sulfonyl chloride
Synonyms: 4-{1,3-dioxo-2-azaspiro[4.4]nonan-2-yl}benzenesulfonyl chloride
Molecular Weight (g/mol): 327.78
Molecular Formula: C14H14ClNO4S
InChi Key: MZHDMPSNJZWYSL-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)N1C(=O)CC2(CCCC2)C1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results