accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NHN

CAS RN 13788-84-6

IUPAC Name: 5-methyl-4-phenyl-1H-pyrazole
Synonyms: 3-methyl-4-phenyl-2H-pyrazole
Molecular Weight (g/mol): 158.2
Molecular Formula: C10H10N2
InChi Key: XTXZCNATVCIKTR-UHFFFAOYSA-N
SMILES: CC1=C(C=NN1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)