accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13798-24-8

IUPAC Name: terbium(3+) hexahydrate trichloride
Synonyms: terbium(III) chloride
Molecular Weight (g/mol): 373.37
Molecular Formula: Cl3H12O6Tb
InChi Key: ULJUVCOAZNLCJZ-UHFFFAOYSA-K
SMILES: O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Tb+3]
Physical Form 
  • (2)
  • (4)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
  • (2)
Grade 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)