accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH2NSNH2OO

CAS RN 138-37-4

IUPAC Name: hydrogen 4-(aminomethyl)benzene-1-sulfonamide chloride
Synonyms: hydrogen emilene chloride
Molecular Weight (g/mol): 222.69
Molecular Formula: C7H11ClN2O2S
InChi Key: SIACJRVYIPXFKS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NCC1=CC=C(C=C1)S(N)(=O)=O
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results