accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SOOOHO

CAS RN 138-41-0

IUPAC Name: 4-sulfamoylbenzoic acid
Synonyms: 4-aminosulfonylbenzoic acid
Molecular Weight (g/mol): 201.2
Molecular Formula: C7H7NO4S
InChi Key: UCAGLBKTLXCODC-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results