accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 138-52-3

IUPAC Name: (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
Synonyms: salicin D-salicin delta-salicin D-(-)-salicin salicine salicin (6CI,8CI) salicoside salicyl alcohol glucoside saligenin-β-D-glucopyranoside saligenin β-D-glucopyranoside
Molecular Weight (g/mol): 286.28
Molecular Formula: C13H18O7
InChi Key: NGFMICBWJRZIBI-UJPOAAIJSA-N
SMILES: OC[C@H]1O[C@@H](OC2=CC=CC=C2CO)[C@H](O)[C@@H](O)[C@@H]1O
Quantity 
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (10)
Percent Purity 
  • (3)
  • (3)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (4)
Grade 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (3)
  • (3)
  • (10)
  • (3)
  • (3)
  • (1)
  • (1)
  • (3)
  • (4)
  • (1)
  • (3)