accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OH2HgOONa

CAS RN 138-85-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 361.7
Molecular Formula: C7H6HgNaO3
InChi Key: BERBQUNVJGVZCX-UHFFFAOYSA-M
SMILES: O.[Na+].[O-]C(=O)C1=CC=C([Hg])C=C1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results