accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH2CH3

CAS RN 138-86-3

IUPAC Name: 1-methyl-4-(prop-1-en-2-yl)cyclohex-1-ene
Synonyms: limonene, (+)- 1,8-p-menthadiene D,L-limonene 1,8(9)-p-menthadiene S-(-)-limonene polydipentene dl-limonene (racemic) p-mentha-1,8-diene (dipentene) terpenes and terpenoids
Molecular Weight (g/mol): 136.24
Molecular Formula: C10H16
InChi Key: XMGQYMWWDOXHJM-UHFFFAOYNA-N
SMILES: CC(=C)C1CCC(C)=CC1
Molecular Weight (g/mol) 
  • (5)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Concentration 
  • (2)
Solvent or Matrix 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)