accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNCH3NO

CAS RN 138-89-6

IUPAC Name: N,N-dimethyl-4-nitrosoaniline
Synonyms: NDMA p-nitrosodimethylanilide paranitrosodimethylanilide P-nitroso-N,N-dimethylaniline
Molecular Weight (g/mol): 150.18
Molecular Formula: C8H10N2O
InChi Key: CMEWLCATCRTSGF-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(C=C1)N=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)