accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13813-40-6

IIITb

CAS RN 13813-40-6

IUPAC Name: terbium(3+) triiodide
Synonyms: terbium(III) iodide
Molecular Weight (g/mol): 539.64
Molecular Formula: I3Tb
InChi Key: OJXRJPFRTRETRN-UHFFFAOYSA-K
SMILES: [I-].[I-].[I-].[Tb+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results