accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13814-96-5

BFFFFBFFFFPb

CAS RN 13814-96-5

IUPAC Name: λ²-lead(2+) bis(tetrafluoroboranuide)
Synonyms: λ²-lead(2+) ditetrafluoroborate
Molecular Weight (g/mol): 380.8
Molecular Formula: B2F8Pb
InChi Key: UQGXAENNKXXYNF-UHFFFAOYSA-N
SMILES: [Pb++].F[B-](F)(F)F.F[B-](F)(F)F
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results