accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHO

CAS RN 13826-35-2

IUPAC Name: (3-phenoxyphenyl)methanol
Synonyms: 3-phenoxybenzyl alcohol 3-pboh
Molecular Weight (g/mol): 200.24
Molecular Formula: C13H12O2
InChi Key: KGANAERDZBAECK-UHFFFAOYSA-N
SMILES: OCC1=CC=CC(OC2=CC=CC=C2)=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)