accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13826-66-9

IUPAC Name: bis(nitric acid); oxozirconium
Synonyms: bis(nitric acid); oxozirconium
Molecular Weight (g/mol): 233.25
Molecular Formula: H2N2O7Zr
InChi Key: UJVRJBAUJYZFIX-UHFFFAOYSA-N
SMILES: O=[Zr].O[N+]([O-])=O.O[N+]([O-])=O
Grade 
  • (6)
Solvent or Matrix 
  • (4)
  • (2)
Technique 
  • (6)
  • (6)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results