accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13826-86-3

NOBFFFFO

CAS RN 13826-86-3

IUPAC Name: nitrous acid; tetrafluoroboranuide
Synonyms: nitronium tetrafluoroborate
Molecular Weight (g/mol): 133.82
Molecular Formula: BF4HNO2
InChi Key: MSXLXAUEYQPTTG-UHFFFAOYSA-N
SMILES: ON=O.F[B-](F)(F)F
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (3)
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (7)
  • (6)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (4)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (2)
  • (3)