accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFOFFFCl

CAS RN 13838-16-9

IUPAC Name: 2-chloro-1-(difluoromethoxy)-1,1,2-trifluoroethane
Synonyms: enflurane enlirane efrane pisa brand of enflurane methylflurether etran baxter anaesthesia brand of enflurane zeneca brand of enflurane enflurane preparation ethrane
Molecular Weight (g/mol): 184.49
Molecular Formula: C3H2ClF5O
InChi Key: JPGQOUSTVILISH-UHFFFAOYNA-N
SMILES: FC(F)OC(F)(F)C(F)Cl
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)