accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13845-07-3

RhClClClClClClKKK

CAS RN 13845-07-3

IUPAC Name: rhodium(3+) tripotassium hexachloride
Synonyms: rhodium(3+) tripotassium hexachloride
Molecular Weight (g/mol): 432.9
Molecular Formula: Cl6K3Rh
InChi Key: OTOHACXAQUCHJO-UHFFFAOYSA-H
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[K+].[K+].[K+].[Rh+3]
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results