accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CdCdOHPOHOHOOHPOHOHOOHPOHOHO

CAS RN 13847-17-1

IUPAC Name: tris(phosphoric acid) dicadmium
Synonyms: tris(phosphoric acid) dicadmium
Molecular Weight (g/mol): 518.81
Molecular Formula: Cd2H9O12P3
InChi Key: GADIEZXPVDTBSM-UHFFFAOYSA-N
SMILES: [Cd].[Cd].OP(O)(O)=O.OP(O)(O)=O.OP(O)(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results