accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CH3COBOCH3H3CNOO

CAS RN 138500-87-5

IUPAC Name: 2-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-{[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}isoindole-1,3-dione
Molecular Weight (g/mol): 363.22
Molecular Formula: C21H22BNO4
InChi Key: ZLSIUDAKDAXZRV-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C(CN2C(=O)C3=CC=CC=C3C2=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results