accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 138605-00-2

ClH3CNCH3OO(S)(S)OH3CO(R)CH3O(R)CH3O(S)OH(R)CH3(S)OHH3CH3C(S)CH2H

CAS RN 138605-00-2

IUPAC Name: hydrogen (3R,4aR,5S,6S,6aS,10S,10aR,10bS)-5-(acetyloxy)-3-ethenyl-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-dodecahydro-1H-naphtho[2,1-b]pyran-6-yl 3-(dimethylamino)propanoate chloride
Synonyms: colforsin
Molecular Weight (g/mol): 546.1
Molecular Formula: C27H44ClNO8
InChi Key: VIRRLEDAYYYTOD-YHEOSNBFSA-N
SMILES: [H+].[Cl-].CN(C)CCC(=O)O[C@@H]1[C@H](OC(C)=O)[C@@]2(C)O[C@](C)(CC(=O)[C@]2(O)[C@@]2(C)[C@@H](O)CCC(C)(C)[C@H]12)C=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results