accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3CH3NNOOHOPOOOH3CHNONNNNH3CONH2CH3CH3OH2NCH3ONH2CH3OH2NH3COH2NH3COH2NH3CHOCH2ONNNNNH2HOHOCo(E/Z)(E/Z)(E/Z)

CAS RN 13870-90-1

IUPAC Name: cobalt(3+) 4,9,14-tris(2-carbamoylethyl)-3,8,19-tris(carbamoylmethyl)-18-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-2,3,6,8,13,13,16,18-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide [5-(6-amino-9H-purin-9-yl)-3,4-dihydroxyoxolan-2-yl]methanide
Synonyms: cobalt(3+) 4,9,14-tris(2-carbamoylethyl)-3,8,19-tris(carbamoylmethyl)-18-{2-[(2-{[5-(5,6-dimethyl-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-2,3,6,8,13,13,16,18-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide [5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methanide
Molecular Weight (g/mol): 1579.61
Molecular Formula: C72H100CoN18O17P
InChi Key: OAJLVMGLJZXSGX-UHFFFAOYNA-L
SMILES: [Co+3].[CH2-]C1OC(C(O)C1O)N1C=NC2=C(N)N=CN=C12.CC(CNC(=O)CCC1(C)C(CC(N)=O)C2[N-]C1=C(C)C1=NC(=CC3=NC(=C(C)C4=NC2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)