CAS RN 138825-96-4
IUPAC Name:
methyl 5-bromo-2-acetamidobenzoate
Synonyms:
methyl 5-bromo-2-acetamidobenzoate
Molecular Weight (g/mol):
272.1
Molecular Formula:
C10H10BrNO3
InChi Key:
CPARHIBNDSEJGR-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC(Br)=CC=C1NC(C)=O
1
–
2
of
2
results
1
Methyl 2-acetamido-5-bromobenzoate, 97%, Thermo Scientific™
CAS: 138825-96-4 Molecular Formula: C10H10BrNO3 Molecular Weight (g/mol): 272.098 MDL Number: MFCD00144758 InChI Key: CPARHIBNDSEJGR-UHFFFAOYSA-N Synonym: 2-acetamido-5-bromobenzoic acid methyl ester,methyl 2-acetylamino-5-bromobenzoate,n-acetyl-5-bromoanthranilic acid methyl ester,benzoic acid, 2-acetylamino-5-bromo-, methyl ester,methyl 5-bromo-2-acetamidobenzoate,acmc-20akiv,pubchem3334,methyl2-acetamido-5-bromobenzoate,labotest-bb lt00454800 PubChem CID: 736223 IUPAC Name: methyl 2-acetamido-5-bromobenzoate SMILES: CC(=O)NC1=C(C=C(C=C1)Br)C(=O)OC