accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNOOOHHO

CAS RN 1389264-18-9

IUPAC Name: 2-oxa-5-azaspiro[3.4]octane; oxalic acid
Synonyms: 2-oxa-5-azaspiro[3.4]octane; oxalic acid
Molecular Weight (g/mol): 203.19
Molecular Formula: C8H13NO5
InChi Key: JFOZNINEJYPQQK-UHFFFAOYSA-N
SMILES: OC(=O)C(O)=O.C1OCC11CCCN1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results