accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClH3CNCH3H3C

CAS RN 139-08-2

IUPAC Name: benzyldimethyltetradecylazanium chloride
Synonyms: miristalkonium chloride
Molecular Weight (g/mol): 368.05
Molecular Formula: C23H42ClN
InChi Key: OCBHHZMJRVXXQK-UHFFFAOYSA-M
SMILES: [Cl-].CCCCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (2)
Industry Type 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results