accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HOONOHOOHO

CAS RN 139-13-9

IUPAC Name: 2-[bis(carboxymethyl)amino]acetic acid
Synonyms: nitrilotriacetic acid acid, nitrilotriacetic hampshire nta acid NTA complexon I trilon A nitrilotriessigsaeure versene nta acid ccris 436 kyselina nitrilotrioctova
Molecular Weight (g/mol): 191.14
Molecular Formula: C6H9NO6
InChi Key: MGFYIUFZLHCRTH-UHFFFAOYSA-N
SMILES: OC(=O)CN(CC(O)=O)CC(O)=O
Quantity 
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
  • (3)
  • (7)
Physical Form 
  • (4)
  • (2)
  • (1)
Grade 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (4)
  • (3)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (7)
  • (4)
  • (2)
  • (1)
  • (1)
  • (2)