CAS RN 1391054-56-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
2-(4,6-Diamino-1,3,5-triazin-2-yl)sulfanylethanesulfonic Acid, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
2-(4,6-Diamino-1,3,5-triazin-2-yl)sulfanylethanesulfonic Acid, TRC
CAS: 1391054-56-0 Molecular Formula: C5 H9 N5 O3 S2 Molecular Weight (g/mol): 251.29 Synonym: 2-[(4,6-Diamino-1,3,5-triazin-2-yl)thio]ethanesulfonic Acid,2-(4,6-Diamino-1,3,5-triazin-2-yl)sulfanylethanesulfonic Acid,Mesna Imp. E (EP) IUPAC Name: 2-[(4,6-diamino-1,3,5-triazin-2-yl)sulfanyl]ethanesulfonic acid SMILES: Nc1nc(N)nc(SCCS(=O)(=O)O)n1