accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FN

CAS RN 13916-99-9

IUPAC Name: 4-fluoronaphthalene-1-carbonitrile
Synonyms: 4-fluoronaphthalene-1-carbonitrile
Molecular Weight (g/mol): 171.17
Molecular Formula: C11H6FN
InChi Key: XQSGYYNFKIQWAC-UHFFFAOYSA-N
SMILES: FC1=CC=C(C#N)C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results