CAS RN 139290-65-6
IUPAC Name:
(2,3-dimethoxyphenyl)({1-[2-(4-fluorophenyl)ethyl]piperidin-4-yl})methanol
Synonyms:
volinanserin
Molecular Weight (g/mol):
373.47
Molecular Formula:
C22H28FNO3
InChi Key:
HXTGXYRHXAGCFP-UHFFFAOYNA-N
SMILES:
COC1=CC=CC(C(O)C2CCN(CCC3=CC=C(F)C=C3)CC2)=C1OC
1
–
3
of
3
results
1
Volinanserin Hydrochloride Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
Volinanserin Hydrochloride Salt, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3