accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PdNNNNClClH2O

CAS RN 13933-31-8

IUPAC Name: dichloropalladium tetraamine hydrate
Synonyms: palladium chloride tetraamine hydrate
Molecular Weight (g/mol): 263.46
Molecular Formula: Cl2H14N4OPd
InChi Key: WVCXSPJPERKPJS-UHFFFAOYSA-L
SMILES: N.N.N.N.O.Cl[Pd]Cl
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results