CAS RN 1393816-99-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Descarbonsildenafil, TRC
CAS: 1393816-99-3 Molecular Formula: C21H30N6O4S Molecular Weight (g/mol): 462.57 Synonym: Benzenesulfonamide, 3-(6,7-dihydro-1-methyl-7-oxo-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-N-[2-(dimethylamino)ethyl]-4-ethoxy-,Descarbonsildenafil,Sildenafil descarbon IUPAC Name: N-[2-(dimethylamino)ethyl]-4-ethoxy-3-(1-methyl-7-oxo-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-5-yl)benzenesulfonamide SMILES: CCCc1nn(C)c2C(=O)NC(=Nc12)c3cc(ccc3OCC)S(=O)(=O)NCCN(C)C