accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

MoCOCOCOCOCOCO

CAS RN 13939-06-5

IUPAC Name: hexakis(methanidylidyneoxidanium) molybdenum
Synonyms: molybdenum hexacarbonyl
Molecular Weight (g/mol): 264.01
Molecular Formula: C6MoO6
InChi Key: KMKBZNSIJQWHJA-UHFFFAOYSA-N
SMILES: [Mo].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
Molecular Weight (g/mol) 
  • (6)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)