CAS RN 139493-40-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,11-Dimethylchrysene, TRC
CAS: 139493-40-6 Molecular Formula: C20H16 Molecular Weight (g/mol): 256.34 SMILES: CC1=CC2=CC=C(C)C=C2C3=C1C(C=CC=C4)=C4C=C3