accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13963-57-0

IUPAC Name: 4-({bis[(4-oxopent-2-en-2-yl)oxy]alumanyl}oxy)pent-3-en-2-one
Synonyms: aluminium acetylacetonate
Molecular Weight (g/mol): 324.31
Molecular Formula: C15H21AlO6
InChi Key: KILURZWTCGSYRE-UHFFFAOYSA-K
SMILES: CC(=O)C=C(C)O[Al](OC(C)=CC(C)=O)OC(C)=CC(C)=O
Boiling Point 
  • (5)
  • (3)
Physical Form 
  • (3)
  • (5)
Quantity 
  • (3)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (3)
  • (5)
  • (3)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)