accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

PPClClPd

CAS RN 13965-03-2

IUPAC Name: palladium(2+) bis(triphenylphosphane) dichloride
Synonyms: bis(triphenylphosphine)palladium chloride
Molecular Weight (g/mol): 701.9
Molecular Formula: C36H30Cl2P2Pd
InChi Key: YNHIGQDRGKUECZ-UHFFFAOYSA-L
SMILES: [Cl-].[Cl-].[Pd++].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (13)
Quantity 
  • (4)
  • (1)
  • (1)
  • (4)
  • (4)
Physical Form 
  • (3)
  • (4)
  • (6)
Percent Purity 
  • (3)
  • (4)
  • (3)
Grade 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (13)
  • (4)
  • (1)
  • (1)
  • (4)
  • (4)
  • (3)
  • (4)
  • (6)
  • (3)
  • (4)
  • (3)
  • (3)
  • (3)