accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 13966-94-4

InI

CAS RN 13966-94-4

IUPAC Name: λ¹-indiganyliumylidene iodide
Synonyms: λ¹-indiganyliumylidene iodide
Molecular Weight (g/mol): 241.72
Molecular Formula: IIn
InChi Key: FOVZCYAIUZHXGB-UHFFFAOYSA-M
SMILES: [In+].[I-]
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results