CAS RN 13967-50-5
CAS RN 13967-50-5
IUPAC Name:
gold(1+) potassium bis(iminomethanide)
Synonyms:
gold(1+) potassium bis(cyanide)
Molecular Weight (g/mol):
288.1
Molecular Formula:
C2AuKN2
InChi Key:
XTFKWYDMKGAZKK-UHFFFAOYSA-N
SMILES:
[K+].[Au+].[C-]#N.[C-]#N
1
–
2
of
2
results
1
Gold(I) potassium cyanide, Premion™, 99.96% (metals basis), Au 67.6% min
CAS: 13967-50-5 MDL Number: MFCD00011414 Synonym: Potassium dicyanoaurate(I); Potassium gold cyanide
2
Gold(I) potassium cyanide, Premion™, 99.96% (metals basis), Au 67.6% min
Form: Crystalline powder | CAS: 13967-50-5 | C{2}AuKN{2}